Merck
  • Scrub typhus during pregnancy and its treatment: a case series and review of the literature.
Scrub typhus during pregnancy and its treatment: a case series and review of the literature.
The American journal of tropical medicine and hygiene (2006-11-25)
Yeon-Sook Kim, Hyo Jin Lee, Meayoung Chang, Sung Kyong Son, Yun Ee Rhee, Soo Kyong Shim
ABSTRACT
Although scrub typhus is uncommon in pregnant women, when present, it can have serious repercussions for the mother and developing fetus. Since it is uncommon, the clinical impact of scrub typhus on pregnancy has not been elucidated and an effective and safe therapeutic regimen has not been validated. The medical records of pregnant women whose scrub typhus were treated at Chungnam National University Hospital were reviewed and their clinical outcomes were evaluated. A review of the literature was also performed on pregnant women with scrub typhus and their clinical outcomes. Eight pregnant women with scrub typhus were treated successfully with a single 500-mg dose of azithromycin, and no relapses were reported. They all delivered healthy babies at term, without congenital or neonatal complications. In the reviews, azithromycin was effective against scrub typhus and had favorable pregnancy outcomes. Ciprofloxacin and cefuroxime failed to treat scrub typhus and fetal loss resulted. A single 500-mg dose of azithromycin may be a reasonable treatment regimen for pregnant women with scrub typhus. Ciprofloxacin might not be advisable for the treatment of scrub typhus during pregnancy. Scrub typhus itself seems to have serious adverse effects on pregnancy if not appropriately controlled.
Product Description
Supelco
Clencyclohexerol hydrochloride, VETRANAL®, analytical standard
Pricing and availability is not currently available.