Merck
  • Albendazole for the control and elimination of lymphatic filariasis: systematic review.
Albendazole for the control and elimination of lymphatic filariasis: systematic review.
Tropical medicine & international health : TM & IH (2005-09-02)
Julia Critchley, David Addiss, Henry Ejere, Carrol Gamble, Paul Garner, Hellen Gelband
ABSTRACT
The Global Programme to Eliminate Lymphatic Filariasis recommends albendazole in combination with other antifilarial drugs. This systematic review examines albendazole in treatment and control of lymphatic filariasis. DATASOURCES: The Cochrane Controlled Trials Register, MEDLINE and EMBASE to April 2005; contacting experts, international organisations and drug manufacturers. Randomised or quasi-randomised controlled trials included; two reviewers independently assessed eligibility, quality, and extracted data. We calculated the relative risk of microfilaraemia (mf) prevalence using fixed effect, or random effects model in case of heterogeneity. Six trials met inclusion criteria. Three trials compared albendazole with placebo: no effect was demonstrated on mf prevalence, but density was lower in one of the three studies at 6 months. Three trials added albendazole to ivermectin, with no demonstrable effect; prevalence tended to be lower at 4--6 months but not at 12 months (4--6 months; RR 0.49, 95% CI 0.18 to 1.39, n=255, 2 trials; 12 months: RR 1.00, 95% CI 0.88 to 1.13, n=348, 2 trials). Mf density was significantly lower in two of the three trials; one of two trials measuring density at 12 months showed a difference. Three trials added albendazole to diethylcarbamazine; two were small trials with no difference demonstrated; the third study tended to favour combination at 6 months (RR=0.62, 95% CI 0.32 to 1.21, n=491), with a significant difference for density. The effect of albendazole against adult and larval filarial parasites, alone and in combination with other antifilarial drugs, deserves further rigorous research.
Product Description
Supelco
Albendazole, analytical standard, ≥98%
Albendazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Albendazole, Pharmaceutical Secondary Standard; Certified Reference Material