Merck
  • Efficacy of indapamide 1.5 mg, sustained release, in the lowering of systolic blood pressure.
Efficacy of indapamide 1.5 mg, sustained release, in the lowering of systolic blood pressure.
Journal of human hypertension (2004-12-14)
G M London
ABSTRACT
The relationship between the increase in blood pressure and the incidence of cardiovascular disease is well recognized today. Studies have shown that more attention should be paid to systolic blood pressure (SBP) in relation to cardiovascular risk and that therapeutic interventions should preferably focus on reducing SBP. The antihypertensive efficacy of indapamide 1.5 mg sustained release (indapamide SR), a low-dose thiazide-type diuretic, was assessed on SBP. Three randomized, double-blind, controlled studies were conducted with indapamide SR, over a period of 3 to 12 months. Elderly patients or patients with target-organ damage, hypertension and left ventricular hypertrophy (LVH) (LIVE study) or with type II diabetes with microalbuminuria (NESTOR study) showed a decrease in SBP varying from 22.7 to 31.8 mmHg. The treatment with indapamide SR resulted in a better or equivalent control of SBP than treatment with a standard dose of a true thiazide diuretic (hydrochlorothiazide), a calcium channel blocker (amlodipine), and an angiotensin-converting enzyme inhibitor (enalapril). No therapeutic escape was observed. All treatments showed good acceptability with no unexpected adverse event. In conclusion, indapamide SR is very effective in lowering SBP-a major independent cardiovascular risk factor-notably in hypertensive high-risk patients with LVH, the elderly and diabetics, when compared to major antihypertensive treatments. This SBP-lowering effect is maintained over the long term.
Product Description
Supelco
Indapamide, analytical standard, for drug analysis