Merck
  • Abuse and dependence potential for the non-benzodiazepine hypnotics zolpidem and zopiclone: a review of case reports and epidemiological data.
Abuse and dependence potential for the non-benzodiazepine hypnotics zolpidem and zopiclone: a review of case reports and epidemiological data.
Addiction (Abingdon, England) (2003-10-02)
G Hajak, W E Müller, H U Wittchen, D Pittrow, W Kirch
ABSTRACT
The non-benzodiazepine hypnotics zolpidem and zopiclone, which are indicated for short-term treatment of insomnia, were considered originally by physicians as almost devoid of abuse and dependence potential. Several recent publications, however, have suggested that both agents carry a significant risk of abuse. To substantiate and re-evaluate this situation, the world literature was reviewed for cases of dependence of both agents; these cases were analysed in order to identify certain underlying patterns, if evident. A systematic review based on a Medline literature search was conducted including the years 1966-2002 to assemble all available clinical case reports that were analysed for typical features of abuse and dependence according to prespecified criteria. Only case reports were of interest, and clinical studies were excluded. No limitations as to language or publication year were applied. The terms 'zolpidem', 'zopiclone' and 'abuse', 'dependence', 'addiction', 'withdrawal' and 'intoxication' were used to identify relevant publications. Potentially relevant citations were retrieved and assessed for inclusion independently by two authors. A total of 36 cases for zolpidem were identified, most of them reported in recent years, and 22 cases for zopiclone. Both sexes were involved to a similar extent; and cases were reported in all age groups. In extreme cases, dose increases reached a factor of 30-120 above the recommended doses. The majority of patients had a history of former drug or alcohol abuse and/or other psychiatric conditions. On the basis of world-wide prescription numbers, which are approximately twofold higher for zolpidem (1,338,774,000 tablets from June 2001 to June 2002 in Europe, Japan and United States) than for zopiclone (664,897,000 tablets during the same period in Europe and Japan), the relative incidence of reported dependence similar for both drugs and remarkably lower than that of benzodiazepines used for the treatment of disturbed sleep. The findings offer the conclusion that zolpidem and zopiclone are relatively safe drugs. However, as both drugs are psychotropic drugs, patients with a history of abuse or dependence and those with psychiatric diseases seem to be at increased risk of abuse of these agents.
Product Description
Supelco
Zopiclone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®