Merck
Oleamide: a member of the endocannabinoid family?
British journal of pharmacology (2003-12-24)
Christopher J Fowler
ABSTRACT
The fatty acid amide class of compounds, which include the endocannabinoid anandamide and the sleep-inducing compound oleamide, have been shown in vitro to have a multiplicity of actions upon different neurochemical systems. In the present issue of this journal, Leggett et al present data indicating that oleamide functionally activates CB(1) cannabinoid receptors in vitro. The significance of their finding is discussed in this commentary.
Sigma-Aldrich
Oleamide, ≥99%
Pricing and availability is not currently available.
Supelco
Oleamide, analytical standard
Pricing and availability is not currently available.
Plastic additive 20, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.