Merck
  • Features of mometasone furoate nasal spray and its utility in the management of allergic rhinitis.
Features of mometasone furoate nasal spray and its utility in the management of allergic rhinitis.
Expert opinion on pharmacotherapy (2003-08-29)
Eric Schenkel
ABSTRACT
Mometasone furoate aqueous nasal spray (NS; Nasonex, Schering Corporation), is a synthetic corticosteroid approved for the prophylaxis and treatment of seasonal allergic rhinitis (SAR) and the treatment of perennial allergic rhinitis (PAR) in patients >or= 12 years of age, and for the treatment of SAR and PAR in children as young as 2 years of age. Studies demonstrate that mometasone furoate NS is a potent, clinically effective and well-tolerated intranasal corticosteroid with negligible systemic activity and which offers the convenience of once-daily dosing.
Product Description
Supelco
Mometasone Furoate, Pharmaceutical Secondary Standard; Certified Reference Material
Mometasone furoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mometasone furoate, ≥98% (HPLC)