Merck
Salicylic acid: an old dog, new tricks, and staphylococcal disease.
The Journal of clinical investigation (2003-07-17)
Mathias Herrmann
ABSTRACT
Aspirin has been shown to cause a reduction in the virulence of Staphylococcus aureus-associated endocarditis. A new study reveals that salicylic acid, the major metabolite of aspirin, acts at the level of transcription to downregulate the production of fibrinogen, fibronectin, and alpha-hemolysin - virulence factors necessary for bacterial replication in host tissues and, now, potential therapeutic targets.
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®