Merck
  • On the mammalian acetone metabolism: from chemistry to clinical implications.
On the mammalian acetone metabolism: from chemistry to clinical implications.
Biochimica et biophysica acta (2003-05-03)
Miklós Péter Kalapos
ABSTRACT
Despite the description of the ways of acetone metabolism, its real role(s) is (are) still unknown in metabolic network. In this article, a trial is made to ascertain a comprehensive overview of acetone research extending discussion from chemistry to clinical implications. Mammals are quite similar regarding their acetone metabolism, even if species differences can also be observed. By reviewing experimental data, it seems that plasma concentration of acetone in different species is in the order of 10 microm range and the concentration-dependent acetone metabolism is common to all mammals. At low concentrations of plasma acetone, the C3 pathways are operative, while at higher concentrations, the metabolism through acetate becomes dominant. Glucose formation from acetone may also contribute to the maintenance of a constant blood glucose level, but it seems to be only a minor source for that. From energetical point of view, an interorgan cooperation is suggested because transportable C3 fragments produced in the liver can serve as alternative sources of energy for the peripheral tissues in the short of circulating glucose. The degradation of acetoacetate to acetone contributes to the maintenance of pH buffering capacity, as well. Special attention is paid to the discussion of acetone production in diseases amongst which endogenous and exogenous acetonemiae have been defined. Acetonemiae of endogenous origin are due to the increased rate of acetone production followed by an increase of degrading capacity as cytochrome p450IIE1 (CYPIIE1) isozymes become induced. Exogenous acetonemiae usually resulted from intoxications caused by either acetone itself or other exogenous compounds (ethanol, isopropyl alcohol). It is highlighted that, on the one hand, isopropanol is also a normal constituent of metabolism and, on the other hand, the flat opinion that the elevation of its plasma level is a sign of alcoholism cannot further be held. The possible future directions of research upon acetone are depicted by emphasizing the need for the clear-cut identification of mammalian acetoacetate decarboxylase, and the investigation of race differences and genetic background of acetone metabolism.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99.5%, ACS reagent
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%