Merck
  • Solubility prediction of salmeterol xinafoate in water--dioxane mixtures.
Solubility prediction of salmeterol xinafoate in water--dioxane mixtures.
International journal of pharmaceutics (2001-03-29)
A Jouyban-Gharamaleki, P York, M Hanna, B J Clark
ABSTRACT
The mole fraction solubility of salmeterol xinafoate was determined in various concentrations of dioxane in aqueous binary mixture. Maximum solubility was observed in 90% v/v dioxane and solubility parameter of the solute was estimated from solubility peak equal to 24.99 MPa(0.5). The predicting capability of four different cosolvency models was also evaluated employing a five data point training set. The solubility data at other cosolvent concentrations were predicted using the trained models, with percentage average errors for 28 drug solubility data sets in water-cosolvent mixtures lying between 12.5 and 15.0%. Further predictive model is proposed for accurate solubility predictions based on a minimum number of experiments. The percentage average error where tested was 10.6%.
Product Description
Salmeterol xinafoate for system suitability, European Pharmacopoeia (EP) Reference Standard
Salmeterol xinafoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Salmeterol xinafoate, ≥98% (HPLC), solid