Merck
  • Protective effect of genistein on lipopolysaccharide/D-galactosamine- induced hepatic failure in mice.
Protective effect of genistein on lipopolysaccharide/D-galactosamine- induced hepatic failure in mice.
Biological & pharmaceutical bulletin (2014-05-13)
Xing Lin, Shijun Zhang, Renbin Huang, Ling Wei, Chunhong Liang, Yongxing Chen, Shujuan Lv, Shuang Liang, Xiaoyan Wu, Quanfang Huang
ABSTRACT
This study examined the effect of genistein from Hydrocotyle sibthorpioides on lipopolysaccharide (LPS)/D-galactosamine (D-GalN)-induced acute hepatic failure. Compared to the model control, genistein treatment significantly protected against LPS/D-GalN-induced liver injury, as evidenced by the decrease in serum alanine and aspartate aminotransferases activities and the attenuation of histopathological changes. Furthermore, genistein alleviated the pro-inflammatory cytokines including tumor necrosis factor-alpha (TNF-α) and nitric oxide (NO)/inducible nitric oxide synthase (iNOS) by inhibiting nuclear factor-κB (NF- κB) activity. Genistein attenuated the elevated level of caspases-3, while augmented the expression of Bcl-2. In addition, LPS/D-GalN induced significant increase of heme oxygenase (HO), carbon monoxide and bilirubin levels and these alterations were augmented by genistein treatment. In conclusion, the protective effect of genistein on LPS/D-GalN-induced liver damage was mainly due to its ability to block NF-κB signaling pathway for anti-inflammation response, attenuate hepatocellular apoptosis and increase HO level. These findings suggest that genistein can be considered as a potential agent for preventing acute hepatic failure.
Sigma-Aldrich
D-(+)-Galactosamine hydrochloride, ≥99% (HPLC)
Pricing and availability is not currently available.