Merck
  • Determination of kaempferol and quercetin in Xindakang tablet by β-cyclodextrin modified micellar electrokinetic capillary chromatography.
Determination of kaempferol and quercetin in Xindakang tablet by β-cyclodextrin modified micellar electrokinetic capillary chromatography.
Pakistan journal of pharmaceutical sciences (2014-05-09)
Yuanqi Lu, Dunqing Wang, Zhihua Ning, Qingqiang Yao
ABSTRACT
A method was proposed to determine kaempferol and quercetin in Hippophae rhamnoides L medicinal preparation xindakang tablet by β-cyclodextrin modified micellar electrokinetic capillary chromatography. Under the optimized conditions: buffer solution of 20 mmol/L Na(2)B(4)O(7)-KH(2)PO(4) (pH 9.0)-20mmol/L SDS-6mmol/L β-CD-5%(v/v) MeCN, applied voltage of 16 kV and injection time of 8s, the two analytes were separated well within 10 minutes. The calibration was linear in the 0.02-0.80 and 0.02-0.70 mg/mL range for kaempferol and quercetin, respectively. The reproducibility based on migration time and peak height were 0.47% and1.87% for kaempferol, 0.55% and 2.02% for quercetin. The detection limits based on three times noise were 0.010 mg/mL and 0.008 mg/mL for kaempferol and quercetin, respectively. The developed method was utilized to analyze real samples and running recovery experiments with satisfactory results.
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG, ≥98.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG, ≥95.0% (HPLC)
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Supelco
Kaempferol, analytical standard
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Betadex, European Pharmacopoeia (EP) Reference Standard
Supelco
Beta-Cyclodextrin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG
Sigma-Aldrich
β-Cyclodextrin, ≥97%