Merck
  • Treatment of HCV with ABT-450/r-ombitasvir and dasabuvir with ribavirin.
Treatment of HCV with ABT-450/r-ombitasvir and dasabuvir with ribavirin.
The New England journal of medicine (2014-04-12)
Jordan J Feld, Kris V Kowdley, Eoin Coakley, Samuel Sigal, David R Nelson, Darrell Crawford, Ola Weiland, Humberto Aguilar, Junyuan Xiong, Tami Pilot-Matias, Barbara DaSilva-Tillmann, Lois Larsen, Thomas Podsadecki, Barry Bernstein
ABSTRACT
The interferon-free combination of the protease inhibitor ABT-450 with ritonavir (ABT-450/r) and the NS5A inhibitor ombitasvir (also known as ABT-267) plus the nonnucleoside polymerase inhibitor dasabuvir (also known as ABT-333) and ribavirin has shown efficacy against the hepatitis C virus (HCV) in patients with HCV genotype 1 infection. In this phase 3 trial, we evaluated this regimen in previously untreated patients with HCV genotype 1 infection and no cirrhosis. In this multicenter, randomized, double-blind, placebo-controlled trial, we assigned previously untreated patients with HCV genotype 1 infection, in a 3:1 ratio, to an active regimen consisting of a single-tablet coformulation of ABT-450/r-ombitasvir (at a once-daily dose of 150 mg of ABT-450, 100 mg of ritonavir, and 25 mg of ombitasvir), and dasabuvir (250 mg twice daily) with ribavirin (in doses determined according to body weight) (group A) or matching placebos (group B). The patients received the study treatment during a 12-week double-blind period. The primary end point was sustained virologic response at 12 weeks after the end of treatment. The primary analysis compared the response rate in group A with the response rate (78%) in a historical control group of previously untreated patients without cirrhosis who received telaprevir with peginterferon and ribavirin. Adverse events occurring during the double-blind period were compared between group A and group B. A total of 631 patients received at least one dose of the study drugs. The rate of sustained virologic response in group A was 96.2% (95% confidence interval, 94.5 to 97.9), which was superior to the historical control rate. Virologic failure during treatment and relapse after treatment occurred in 0.2% and 1.5%, respectively, of the patients in group A. The response rates in group A were 95.3% among patients with HCV genotype 1a infection and 98.0% among those with HCV genotype 1b infection. The rate of discontinuation due to adverse events was 0.6% in each study group. Nausea, pruritus, insomnia, diarrhea, and asthenia occurred in significantly more patients in group A than in group B (P<0.05 for all comparisons). Reductions in the hemoglobin level were all of grade 1 or 2; reductions of grade 1 and 2 occurred in 47.5% and 5.8%, respectively, of the patients in group A, whereas grade 1 reductions occurred in 2.5% of the patients in group B. In previously untreated patients with HCV genotype 1 infection and no cirrhosis, a 12-week multitargeted regimen of ABT-450/r-ombitasvir and dasabuvir with ribavirin was highly effective and was associated with a low rate of treatment discontinuation. (Funded by AbbVie; SAPPHIRE-I ClinicalTrials.gov number, NCT01716585.).
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Uracil, ≥99.0%
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard