Merck
Erbin is a negative modulator of cardiac hypertrophy.
Proceedings of the National Academy of Sciences of the United States of America (2014-04-09)
Inbal Rachmin, Sagi Tshori, Yoav Smith, Amit Oppenheim, Sylvie Marchetto, Gillian Kay, Roger S-Y Foo, Noa Dagan, Eliahu Golomb, Dan Gilon, Jean-Paul Borg, Ehud Razin
ABSTRACT
ErbB2 interacting protein (Erbin) is a widely expressed protein and participates in inhibition of several intracellular signaling pathways. Its mRNA has been found to be present in relatively high levels in the heart. However, its physiological role in the heart has not been explored. In the present work, we elucidated the role of Erbin in cardiac hypertrophy. Cardiac hypertrophy was induced in mice either by isoproterenol administration or by aortic constriction. The level of Erbin was significantly decreased in both models. Erbin(-/-) mice rapidly develop decompensated cardiac hypertrophy, and following severe pressure overload all Erbin(-/-) mice died from heart failure. Down-regulation of Erbin expression was also observed in biopsies derived from human failing hearts. It is known that Erbin inhibits Ras-mediated activation of the extracellular signal-regulated kinase (ERK) by binding to Soc-2 suppressor of clear homolog (Shoc2). Our data clearly show that ERK phosphorylation is enhanced in the heart tissues of Erbin(-/-) mice. Furthermore, we clearly demonstrate here that Erbin associates with Shoc2 in both whole hearts and in cardiomyocytes, and that in the absence of Erbin, Raf is phosphorylated and binds Shoc2, resulting in ERK phosphorylation. In conclusion, Erbin is an inhibitor of pathological cardiac hypertrophy, and this inhibition is mediated, at least in part, by modulating ERK signaling.
Sigma-Aldrich
Isoprenaline hydrochloride
Isoprenaline sulfate, European Pharmacopoeia (EP) Reference Standard