Merck
  • Utility of an immunotherapy trial in evaluating patients with presumed autoimmune epilepsy.
Utility of an immunotherapy trial in evaluating patients with presumed autoimmune epilepsy.
Neurology (2014-04-08)
M Toledano, J W Britton, A McKeon, C Shin, V A Lennon, A M L Quek, E So, G A Worrell, G D Cascino, C J Klein, T D Lagerlund, E C Wirrell, K C Nickels, S J Pittock
ABSTRACT
To evaluate a trial of immunotherapy as an aid to diagnosis in suspected autoimmune epilepsy. We reviewed the charts of 110 patients seen at our autoimmune neurology clinic with seizures as a chief complaint. Twenty-nine patients met the following inclusion criteria: (1) autoimmune epilepsy suspected based on the presence of ≥ 1 neural autoantibody (n = 23), personal or family history or physical stigmata of autoimmunity, and frequent or medically intractable seizures; and (2) initiated a 6- to 12-week trial of IV methylprednisolone (IVMP), IV immune globulin (IVIg), or both. Patients were defined as responders if there was a 50% or greater reduction in seizure frequency. Eighteen patients (62%) responded, of whom 10 (34%) became seizure-free; 52% improved with the first agent. Of those receiving a second agent after not responding to the first, 43% improved. A favorable response correlated with shorter interval between symptom onset and treatment initiation (median 9.5 vs 22 months; p = 0.048). Responders included 14/16 (87.5%) patients with antibodies to plasma membrane antigens, 2/6 (33%) patients seropositive for glutamic acid decarboxylase 65 antibodies, and 2/6 (33%) patients without detectable antibodies. Of 13 responders followed for more than 6 months after initiating long-term oral immunosuppression, response was sustained in 11 (85%). These retrospective findings justify consideration of a trial of immunotherapy in patients with suspected autoimmune epilepsy. This study provides Class IV evidence that in patients with suspected autoimmune epilepsy, IVMP, IVIg, or both improve seizure control.
Sigma-Aldrich
6α-Methylprednisolone, ≥98%
Pricing and availability is not currently available.
Methylprednisolone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.