Merck
  • Diclofenac protects cultured human corneal epithelial cells against hyperosmolarity and ameliorates corneal surface damage in a rat model of dry eye.
Diclofenac protects cultured human corneal epithelial cells against hyperosmolarity and ameliorates corneal surface damage in a rat model of dry eye.
Investigative ophthalmology & visual science (2014-03-29)
Ryoichi Sawazaki, Tomoaki Ishihara, Shinya Usui, Erika Hayashi, Kayoko Tahara, Tatsuya Hoshino, Akihiro Higuchi, Shigeru Nakamura, Kazuo Tsubota, Tohru Mizushima
ABSTRACT
Dry eye syndrome (DES) is characterized by an increase in tear osmolarity and induction of the expression and nuclear localization of an osmoprotective transcription factor (nuclear factor of activated T-cells 5 [NFAT5]) that plays an important role in providing protection against hyperosmotic tears. In this study, we screened medicines already in clinical use with a view of finding compounds that protect cultured human corneal epithelial cells against hyperosmolarity-induced cell damage. Viable cell number was determined by the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) method and cellular NFAT5 level was measured by immunoblotting. The rat model for DES was developed by removal of the lacrimal glands, with an assessment of corneal surface damage based on levels of fluorescein staining and epithelial apoptosis. Some nonsteroidal anti-inflammatory drugs (NSAIDs), including diclofenac sodium (diclofenac), were identified during the screening procedure. These NSAIDs were able to suppress hyperosmolarity-induced apoptosis and cell growth arrest. In contrast, other NSAIDs, including bromfenac sodium (bromfenac), did not exert such a protective action. Treatment of cells with diclofenac, but not bromfenac, stimulated both the nuclear localization and expression of NFAT5 under hyperosmotic conditions. In the rat model for DES, topical administration of diclofenac (but not bromfenac) to eyes reduced corneal surface damage without affecting the volume of tear fluid. Diclofenac appears to protect cells against hyperosmolarity-induced cell damage and NFAT5 would play an important role in this protective action. The findings reported here may also indicate that the topical administration of diclofenac to eyes may be therapeutically beneficial for DES patients.
Product Description
Diclofenac potassium, European Pharmacopoeia (EP) Reference Standard
Supelco
Diclofenac sodium salt, analytical standard
Sigma-Aldrich
Diclofenac sodium salt
Supelco
Diclofenac sodium salt, Pharmaceutical Secondary Standard; Certified Reference Material
Diclofenac for system suitability, European Pharmacopoeia (EP) Reference Standard
Diclofenac sodium, European Pharmacopoeia (EP) Reference Standard