Merck
  • Fresh frozen plasma resuscitation attenuates platelet dysfunction compared with normal saline in a large animal model of multisystem trauma.
Fresh frozen plasma resuscitation attenuates platelet dysfunction compared with normal saline in a large animal model of multisystem trauma.
The journal of trauma and acute care surgery (2014-03-26)
Martin Sillesen, Pär I Johansson, Lars S Rasmussen, Guang Jin, Cecilie H Jepsen, Ayesha Imam, John O Hwabejire, Danielle Deperalta, Michael Duggan, Marc DeMoya, George C Velmahos, Hasan B Alam
ABSTRACT
Platelet dysfunction following trauma has been identified as an independent predictor of mortality. We hypothesized that fresh frozen plasma (FFP) resuscitation would attenuate platelet dysfunction compared with 0.9% normal saline (NS). Twelve swine were subjected to multisystem trauma (traumatic brain injury, liver injury, rib fracture, and soft tissue injury) with hemorrhagic shock (40% of estimated blood volume). Animals were left in shock (mean arterial pressure, 30-35 mm Hg) for 2 hours followed by resuscitation with three times shed volume NS (n = 6) or one times volume FFP (n = 6) and monitored for 6 hours. Platelet function was assessed by adenosine diphosphate (ADP)-induced platelet aggregation at baseline, after 2 hours of shock following resuscitation, and 6 hours after resuscitation. Fibrinogen levels and markers of platelet activation (transforming growth factor β [TGF-β], sP-Selectin, and CD40L) as well as endothelial injury (intercellular adhesion molecule 1 [ICAM-1], vascular cell adhesion molecule 1 [VCAM-1]) were also assayed. Thromboelastography was used to measure clotting activity. ADP-induced platelet aggregation was significantly higher in the FFP group (46.3 U vs. 25.5 U, p < 0.01) following resuscitation. This was associated with higher fibrinogen levels (202 mg/dL vs. 80 mg/dL, p < 0.01) but lower endothelial activation (VCAM-1, 1.25 ng/mL vs. 3.87 ng/mL, p = 0.05). Other markers did not differ.After 6 hours of observation, ADP-induced platelet aggregation remained higher in the FFP group (53.8 U vs. 37.0 U, p = 0.03) as was fibrinogen levels (229 mg/dL vs. 153 mg/dL, p < 0.01). Endothelial activation was lower (ICAM-1, 21.0 ng/mL vs. 24.4 ng/mL, p = 0.05), whereas TGF-β levels were higher (2,138 pg/mL vs. 1,802 pg/mL, p = 0.03) in the FFP group. Other markers did not differ. Thromboelastography revealed increased clot strength in the FFP group at both postresuscitation time points. Resuscitation with FFP resulted in an immediate and sustained improvement in platelet function and clot strength compared with high-volume NS resuscitation. This was associated with an increase in fibrinogen levels and an attenuation of endothelial activation.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis