Merck
  • Gene expression regulation mediated through reversible m⁶A RNA methylation.
Gene expression regulation mediated through reversible m⁶A RNA methylation.
Nature reviews. Genetics (2014-03-26)
Ye Fu, Dan Dominissini, Gideon Rechavi, Chuan He
ABSTRACT
Cellular RNAs carry diverse chemical modifications that used to be regarded as static and having minor roles in 'fine-tuning' structural and functional properties of RNAs. In this Review, we focus on reversible methylation through the most prevalent mammalian mRNA internal modification, N(6)-methyladenosine (m(6)A). Recent studies have discovered protein 'writers', 'erasers' and 'readers' of this RNA chemical mark, as well as its dynamic deposition on mRNA and other types of nuclear RNA. These findings strongly indicate dynamic regulatory roles that are analogous to the well-known reversible epigenetic modifications of DNA and histone proteins. This reversible RNA methylation adds a new dimension to the developing picture of post-transcriptional regulation of gene expression.
Product Description
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Adenosine
Pricing and availability is not currently available.
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Pricing and availability is not currently available.
Sigma-Aldrich
Adenosine, ≥99%
Pricing and availability is not currently available.
Adenosine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.