Merck
  • Identification of small molecules that support human leukemia stem cell activity ex vivo.
Identification of small molecules that support human leukemia stem cell activity ex vivo.
Nature methods (2014-02-25)
Caroline Pabst, Jana Krosl, Iman Fares, Geneviève Boucher, Réjean Ruel, Anne Marinier, Sébastien Lemieux, Josée Hébert, Guy Sauvageau
ABSTRACT
Leukemic stem cells (LSCs) are considered a major cause of relapse in acute myeloid leukemia (AML). Defining pathways that control LSC self-renewal is crucial for a better understanding of underlying mechanisms and for the development of targeted therapies. However, currently available culture conditions do not prevent spontaneous differentiation of LSCs, which greatly limits the feasibility of cell-based assays. To overcome these constraints we conducted a high-throughput chemical screen and identified small molecules that inhibit differentiation and support LSC activity in vitro. Similar to reports with cord blood stem cells, several of these compounds suppressed the aryl-hydrocarbon receptor (AhR) pathway, which we show to be inactive in vivo and rapidly activated ex vivo in AML cells. We also identified a compound, UM729, that collaborates with AhR suppressors in preventing AML cell differentiation. Together, these findings provide newly defined culture conditions for improved ex vivo culture of primary human AML cells.
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Adenine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Myriocin from Mycelia sterilia, ≥98% (HPLC), powder
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material