Merck
  • Protective effects of acetaminophen on ibuprofen-induced gastric mucosal damage in rats with associated suppression of matrix metalloproteinase.
Protective effects of acetaminophen on ibuprofen-induced gastric mucosal damage in rats with associated suppression of matrix metalloproteinase.
The Journal of pharmacology and experimental therapeutics (2014-02-06)
Eriko Fukushima, Noriyuki Monoi, Shigeo Mikoshiba, Yutaka Hirayama, Tetsushi Serizawa, Kiyo Adachi, Misao Koide, Motoyasu Ohdera, Michiaki Murakoshi, Hisanori Kato
ABSTRACT
Nonsteroidal anti-inflammatory drugs (NSAIDs) are known to cause gastric mucosal damage as a side effect. Acetaminophen, widely used as an analgesic and antipyretic drug, has gastroprotective effects against gastric lesions induced by absolute ethanol and certain NSAIDs. However, the mechanisms that underlie the gastroprotective effects of acetaminophen have not yet been clarified. In the present study, we examined the role and protective mechanism of acetaminophen on ibuprofen-induced gastric damage in rats. Ibuprofen and acetaminophen were administered orally, and the gastric mucosa was macroscopically examined 4 hours later. Acetaminophen decreased ibuprofen-induced gastric damage in a dose-dependent manner. To investigate the mechanisms involved, transcriptome analyses of the ibuprofen-damaged gastric mucosa were performed in the presence and absence of acetaminophen. Ingenuity pathway analysis (IPA) software revealed that acetaminophen suppressed the pathways related to cellular assembly and inflammation, whereas they were highly activated by ibuprofen. On the basis of gene classifications from the IPA Knowledge Base, we identified the following five genes that were related to gastric damage and showed significant changes in gene expression: interleukin-1β (IL-1β), chemokine (C-C motif) ligand 2 (CCL2), matrix metalloproteinase-10 (MMP-10), MMP-13, and FBJ osteosarcoma oncogene (FOS). Expression of these salient genes was confirmed using real-time polymerase chain reaction. The expression of MMP-13 was the most reactive to the treatments, showing strong induction by ibuprofen and suppression by acetaminophen. Moreover, MMP-13 inhibitors decreased ibuprofen-induced gastric damage. In conclusion, these results suggest that acetaminophen decreases ibuprofen-induced gastric mucosal damage and that the suppression of MMP-13 may play an important role in the gastroprotective effects of acetaminophen.
Product Description
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard