Merck
  • Measuring polyethylene wear in total knee arthroplasty by RSA: differences between weight-bearing and non-weight-bearing positioning.
Measuring polyethylene wear in total knee arthroplasty by RSA: differences between weight-bearing and non-weight-bearing positioning.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2014-01-08)
Emiel A van Ijsseldijk, Edward R Valstar, Berend C Stoel, Ruud de Ridder, Rob G H H Nelissen, Bart L Kaptein
ABSTRACT
Measuring the minimum-joint-space-width (mJSW) in total knee arthroplasty (TKA) in Roentgen stereophotogrammetric analysis (RSA) provides valuable information on polyethylene wear, a leading cause for TKA failure. Most existing studies use non-weight-bearing (NWB) patient positioning. The latter may compromise mJSW measurements due to knee laxity with subsequent non-contact between the TKA components. We investigated the difference in mJSW between weight-bearing (WB) and NWB images and the association with mediolateral (ML) knee stability. At one-year follow-up, 23 TKAs were included from an ongoing RSA study, and ML stability was evaluated. For each examination, the mJSW and femoral-tibial contact locations were measured. A linear regression model was used to analyze the association between the mJSW difference (NWB-WB) with the ML stability and contact locations. The mean mJSW difference was 0.28 mm medially and 0.20 mm laterally. Four TKAs had medium (5-9°) and 19 TKAs had high (<5°) ML stability. A higher mJSW difference was found for TKAs with medium stability (0.36 mm, P = 0.01). In conclusion, mJSW measurements in existing (NWB) RSA studies are influenced by knee laxity, but may still provide information on wear progression based on TKA with high ML stability. A direct comparison of mJSW measurements from WB and NWB data is not possible.
Product Description
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Polyethylene (LDPE), ERM®, certified reference material