Merck
  • Anti-thrombotic effect of a novel formula from Corni fructus with malic acid, succinic acid and citric acid.
Anti-thrombotic effect of a novel formula from Corni fructus with malic acid, succinic acid and citric acid.
Phytotherapy research : PTR (2013-08-06)
Qi-Chun Zhang, Yue Zhao, Hui-Min Bian
ABSTRACT
Our previous investigation had confirmed the inhibition of platelet aggregation of a novel Corni fructus-derived formula composed of malic acid, succinic acid and citric acid with a ratio of 3:2:2. The present study was to further evaluate the anti-thrombotic effect of the formula in vivo. Mice of acute pulmonary thromboembolism, and rats of arterial thrombosis were used to determine the anti-thrombotic effect of the formula. Histology analysis of endothelium was conducted with hematoxylin and eosin stain. TXB2 , 6-K-PGF1α , cAMP, cGMP and NO in rat plasma were determined. In vitro assay of αIIbβ3 and phosphorylation of ERK1/2 were performed in ADP-treated platelet. The formula significantly reduced the recovery time and mortality rate of mice with acute pulmonary thromboembolism. Remarkably extended occlusion time, decreased thrombus weight and more integrated endothelium were observed in rat with the formula. Enhanced 6-K-PGF1α , cGMP and NO, but not TXB2 and cAMP, were demonstrated in rat plasma with treatment of the formula. Finally, the formula was shown to inhibit αIIbβ3 expression and activation of ERK1/2 in platelet. The formula shows positive anti-thrombotic effect. The direct interference on ADP activated signaling in platelet and regulation of endothelium function are two primary pathways involved in the action on thrombosis.
Sigma-Aldrich
L-(−)-Malic acid disodium salt, ≥95% (titration)
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Malic acid, analytical standard
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Supelco
Succinic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Supelco
Succinic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
DL-Malic acid, 99%
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Sigma-Aldrich
D-(+)-Malic acid, unnatural form, ≥97.0% (T)
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Supelco
Malic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications
Supelco
DL-Malic acid, analytical standard