Merck
  • Access to new 2-oxofuro[2,3-b]pyrroles and 2-methylenepyrroles through the reaction of 1,2-diaza-1,3-butadienes and gamma-ketoesters.
Access to new 2-oxofuro[2,3-b]pyrroles and 2-methylenepyrroles through the reaction of 1,2-diaza-1,3-butadienes and gamma-ketoesters.
The Journal of organic chemistry (2004-04-13)
Orazio A Attanasi, Lucia De Crescentini, Gianfranco Favi, Paolino Filippone, Fabio Mantellini, Stefania Santeusanio
ABSTRACT
New and interesting 2-oxofuro[2,3-b]pyrroles and 19-methyl-15-oxa-20-azatricyclo[12.3.3.0(1,14)]icos-18-en-18-carboxylates have been obtained in good yields by the one-pot reaction, in basic medium, of 1,2-diaza-1,3-butadienes with diethyl or dimethyl acetylsuccinate or methyl 2-(1,3-dioxo-2-cyclotetradecyl)acetate, respectively, under mild conditions. Treatment of the same starting materials with diethyl 2-acetylglutarate, in acidic medium, afforded unknown 2-methylenepyrrole derivatives in high yields. Novel 4-(3-oxopropyl)-2,5-dimethyl-1H-pyrrole-3-carboxylates also have been achieved by reacting 1,2-diaza-1,3-butadienes with ethyl or methyl 4-acetyl-5-oxo-hexanoate.