Merck
  • Synthesis and antiprotozoal activity of novel bis-benzamidino imidazo[1,2-a]pyridines and 5,6,7,8-tetrahydro-imidazo[1,2-a]pyridines.
Synthesis and antiprotozoal activity of novel bis-benzamidino imidazo[1,2-a]pyridines and 5,6,7,8-tetrahydro-imidazo[1,2-a]pyridines.
Bioorganic & medicinal chemistry (2007-11-03)
Mohamed A Ismail, Reem K Arafa, Tanja Wenzler, Reto Brun, Farial A Tanious, W David Wilson, David W Boykin
ABSTRACT
The key dinitrile intermediates 4a-d were synthesized by reaction of phenacyl bromide 1 and the appropriate 2-amino-5-bromopyridines to yield 3a-d. Suzuki coupling of 3a-d with 4-cyanophenylboronic acid yielded the 2,6-bis(4-cyanophenyl)-imidazo[1,2-a]pyridine derivatives 4a-d. The bis-amidoximes 5a-d, obtained from 4a-d by the action of hydroxylamine, were converted to the bis-O-acetoxyamidoximes which on catalytic hydrogenation in a mixture of ethanol/ethyl acetate gave the acetate salts of 2,6-bis[4-(amidinophenyl)]-imidazo[1,2-a]pyridines 7a-d. In contrast, catalytic hydrogenation of the bis-O-acetoxyamidoxime of 5a in glacial acetic acid gave the saturated analogue 2,6-bis[4-(amidinophenyl)]-5,6,7,8-tetrahydro-imidazo[1,2-a]pyridine 8. O-Methylation of the amidoximes 5a-d gave the N-methoxyamidines 6a-d. The diamidines showed strong DNA binding affinity, were very active in vitro against T. b. r. exhibiting IC(50) values between 7 and 38nM, but were less effective against P. f. with IC(50) values between 23 and 92nM. Two of the diamidines 7c and 7d were slightly more active than furamidine but less active than azafuramidine in the T. b. r. STIB900 mouse model. Only one prodrug 6b showed moderate activity in the same mouse model.
Sigma-Aldrich
Imidazo[1,2-a]pyridine, 99%