Merck
  • Effect of hydrophobic octadecyl groups on pH-sensitive aggregation behavior of imidazole-containing polyaspartammide derivatives.
Effect of hydrophobic octadecyl groups on pH-sensitive aggregation behavior of imidazole-containing polyaspartammide derivatives.
Journal of nanoscience and nanotechnology (2010-12-09)
Kwangwon Seo, Doo Sung Lee, Sung Chul Kim, Dukjoon Kim
ABSTRACT
The effect of hydrophobic octadecyl groups on pH-dependent aggregation behavior of polyaspartammide derivatives was studied. A series of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution were synthesized through a successive graft reaction of octaceylamine, O-(2-aminoethyl)-O'-methylpolyethylene glycol, and 1-(3-aminopropyl)imidazole on polysuccinimide. Micelle-like nano-aggregates were formed in aqueous solution at pH > 6.8 and they showed different pH dependent aggregation behavior according to degree of substitution (DS) of the hydrophobic C18 chains. These polymers will have a potential application as carriers for pH-sensitive drug release in anticancer or intracellular delivery systems.
Sigma-Aldrich
1-(3-Aminopropyl)imidazole, ≥97%
Pricing and availability is not currently available.