Merck
  • The precursor strategy: terminus methoxyoxalamido modifiers for single and multiple functionalization of oligodeoxyribonucleotides.
The precursor strategy: terminus methoxyoxalamido modifiers for single and multiple functionalization of oligodeoxyribonucleotides.
Nucleic acids research (2000-08-10)
N N Polushin
ABSTRACT
Synthesis of new terminus modifiers, bearing, along with a phosphoramidite moiety, one, two or four methoxyoxalamido (MOX) precursor groups, is described. These modifiers are introduced onto the 5'-end of a synthetic oligodeoxyribonucleotide as the last step of an automated synthesis to form the MOX precursor oligonucleotide. The MOX groups are then post-synthetically derivatized with an appropriate primary amine to construct a 5'-modified oligonucleotide. The efficiency and simplicity of the novel modifying strategy were demonstrated in the synthesis of a number of 5'-functionalized oligonucleotides.
Sigma-Aldrich
trans-4-Aminocyclohexanol hydrochloride, 97%
Pricing and availability is not currently available.