Merck
  • Non-covalent interactions between iodo-perfluorocarbons and hydrogen bond acceptors.
Non-covalent interactions between iodo-perfluorocarbons and hydrogen bond acceptors.
Chemical communications (Cambridge, England) (2009-04-01)
Rafel Cabot, Christopher A Hunter
ABSTRACT
Quantitative studies of the 1 : 1 complexes formed between perfluorohexyl iodide and a variety of hydrogen-bond acceptors have been used to probe the relationship between halogen bonding, hydrogen bonding, desolvation and the electrostatics of non-covalent interactions.
Sigma-Aldrich
Perfluorohexyl iodide, 99%