Merck
  • HPLC and solubility study of the interaction between pindolol and cyclodextrins.
HPLC and solubility study of the interaction between pindolol and cyclodextrins.
Journal of pharmaceutical and biomedical analysis (2005-03-03)
Carmen Gazpio, Miguel Sánchez, Iñigo X García-Zubiri, Itziar Vélaz, Cristina Martínez-Ohárriz, Carmen Martín, Arantza Zornoza
ABSTRACT
The complexation with beta-cyclodextrin (beta-CD) has been investigated using reversed-phase liquid chromatography. The compounds tested have been pindolol and, for comparison purposes, indole and 4-methoxyindole. The retention behaviour has been analysed on a Kromasil 100 C18 column and the mobile phase used was methanol-pH 6 phosphate buffer (15/85v/v) in which beta-CD was incorporated as a mobile phase additive. The decrease in the retention times with increasing concentrations of beta-CD enables the determination of the apparent stability constants of the complexes. In addition, the low solubility of pindolol, a weak base, in pH 12 aqueous solution has been improved by complexation with different cyclodextrins. The solubility enhancements with 1.4 x 10(-2) M beta-, hydroxypropyl-beta, and gamma-CD have been 1.9, 1.8 and 1.4-fold, respectively, with 2.4 x 10(-2) M methyl-beta-CD it was 2.8-fold whilst no effect was observed with alpha-CD. The stability constants of the complexes at pH 12 have been determined from the solubility isotherms.
Sigma-Aldrich
4-Methoxyindole, 99%
Pricing and availability is not currently available.