Merck
  • Xylanase supplementation of a wheat-based diet improved nutrient digestion and mRNA expression of intestinal nutrient transporters in broiler chickens infected with Clostridium perfringens.
Xylanase supplementation of a wheat-based diet improved nutrient digestion and mRNA expression of intestinal nutrient transporters in broiler chickens infected with Clostridium perfringens.
Poultry science (2014-02-27)
Shuangshuang Guo, Dan Liu, Xu Zhao, Changwu Li, Yuming Guo
ABSTRACT
Necrotic enteritis caused by Clostridium perfringens has become prevalent in the European Union due to the withdrawal of antibiotics in poultry feed. In an experiment with a 2 × 2 factorial arrangement, 336 one-day-old male broiler chicks (Ross 308) were assigned to 4 groups with or without C. perfringens challenge and fed wheat-based diets supplemented with or without xylanase at 5,500 U/kg of diet. The study aimed to investigate effects of xylanase addition on growth performance as well as nutrient digestion and absorption of C. perfringens-infected broilers. Before challenge (d 0-14), xylanase-supplemented birds had greater ADG and lower feed conversion ratio (FCR; P < 0.05). During infection (d 14-21), challenge tended to decrease ADG (P = 0.063) and significantly increased FCR (P < 0.05), whereas xylanase addition greatly reduced FCR (P < 0.05). Clostridium perfringens infection decreased AME values and apparent ileal digestibility of DM of diets (P < 0.05). Xylanase supplementation increased AME values regardless of infection and apparent ileal digestibility of CP in challenged birds (P < 0.05). Activities of duodenal α-amylase and chymotrypsin and pancreatic trypsin were decreased by C. perfringens infection (P < 0.05). Xylanase supplementation elevated pancreatic chymotrypsin activity and reduced duodenal α-amylase and trypsin activities (P < 0.05). It also decreased jejunal α-amylase activity and increased pancreatic α-amylase as well as jejunal sucrase activities in uninfected birds (P < 0.05). The duodenal mRNA expression of sodium glucose cotransporter 1 (SGLT1), H(+)-dependent peptide transporter 1 (PepT1), and liver fatty acid-binding protein (L-FABP) were downregulated (P < 0.05), but ileal SGLT1 gene expression was increased by infection (P < 0.05). Xylanase addition upregulated expression of jejunal SGLT1, PepT1, and L-FABP genes as well as ileal PepT1 and L-FABP genes in challenged broilers (P < 0.05). In conclusion, xylanase supplementation of wheat-based diets improved FCR and AME in birds irrespective of C. perfringens infection and elevated apparent ileal digestibility of CP and mRNA expression of nutrient transporters in challenged birds.
Sigma-Aldrich
Xylanase from Trichoderma viride, lyophilized powder, 100-300 units/mg protein
Pricing and availability is not currently available.