Merck
  • Structural analysis of the novel influenza A (H7N9) viral Neuraminidase interactions with current approved neuraminidase inhibitors Oseltamivir, Zanamivir, and Peramivir in the presence of mutation R289K.
Structural analysis of the novel influenza A (H7N9) viral Neuraminidase interactions with current approved neuraminidase inhibitors Oseltamivir, Zanamivir, and Peramivir in the presence of mutation R289K.
BMC bioinformatics (2014-02-26)
Chinh Tran-To Su, Xuchang Ouyang, Jie Zheng, Chee-Keong Kwoh
ABSTRACT
Since late March 2013, there has been another global health concern with a sudden wave of flu infections by a novel strain of avian influenza A (H7N9) virus in China. To-date, there have been more than 100 infections with 23 deaths. It is more worrying as this viral strain has never been detected in humans and only been found to be of low-pathogenicity. Currently, there are 3 effective neuraminidase inhibitors for this H7N9 virus strain, i.e. oseltamivir, zanamivir, and peramivir. These drugs have been used for treatment of the H7N9 influenza in China. However, how these inhibitors work and affect the binding cavity of the novel H7N9 neuraminidase in the presence of potential mutations has not been disclosed. In our study, we investigate steric effects and subsequently show the conformational restraints of the inhibitor-binding site of the non-mutated and mutated H7N9 neuraminidase structures to different drug compounds. Combination of molecular docking and Molecular Dynamics simulation reveal that zanamivir forms more favorable and stable complex than oseltamivir and peramivir when binding to the active site of the H7N9 neuraminidase. And it is likely that the novel influenza A (H7N9) virus adopts a higher probability to acquire resistance to peramivir than the other two inhibitors. Conformational changes induced by the mutation R289K causes loss of number of hydrogen bonds between the inhibitors and the H7N9 viral neuraminidase in 2 out of 3 complexes. In addition, our results of binding-affinity relationships of the 3 inhibitors with the viral neuraminidase proteins of previous pandemics (H1N1, H5N1) and the current novel H7N9 reflected the extent of binding effectiveness of the 3 inhibitors to the novel H7N9 neuraminidase. The results are novel and specific for the A/Hangzhou/1/2013(H7N9) influenza strain. Furthermore, the protocol could be useful for further drug-binding analysis and prediction of future viral mutations to which the virus evolves through adaptation and acquires resistance to the current available drugs.
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein