Merck
  • Different hydrolases involved in bioactivation of prodrug-type angiotensin receptor blockers: carboxymethylenebutenolidase and carboxylesterase 1.
Different hydrolases involved in bioactivation of prodrug-type angiotensin receptor blockers: carboxymethylenebutenolidase and carboxylesterase 1.
Drug metabolism and disposition: the biological fate of chemicals (2013-08-16)
Tomoko Ishizuka, Yasushi Yoshigae, Nobuyuki Murayama, Takashi Izumi
ABSTRACT
Olmesartan medoxomil (OM) is a prodrug-type angiotensin II type 1 receptor blocker (ARB). We recently identified carboxymethylenebutenolidase homolog (CMBL) as the responsible enzyme for OM bioactivation in humans. In the present study, we compared the bioactivating properties of OM with those of other prodrug-type ARBs, candesartan cilexetil (CC) and azilsartan medoxomil (AM), by focusing on interspecies differences and tissue specificity. In in-vitro experiments with pooled tissue subcellular fractions of mice, rats, monkeys, dogs, and humans, substantial OM-hydrolase activities were observed in cytosols of the liver, intestine, and kidney in all the species tested except for dog intestine, which showed negligible activity, whereas lung cytosols showed relatively low activities compared with the other tissues. AM-hydrolase activities were well correlated with the OM-hydrolase activities. In contrast, liver microsomes exhibited the highest CC-hydrolase activity among various tissue subcellular fractions in all the species tested. As a result of Western blot analysis with the tissue subcellular fractions, the band intensities stained with anti-human CMBL and carboxylesterase 1 (CES1) antibodies well reflected OM- and AM-hydrolase activities and CC-hydrolase activity, respectively, in animals and humans. Recombinant human CMBL and CES1 showed significant AM- and CC-hydrolase activities, respectively, whereas CC hydrolysis was hardly catalyzed with recombinant carboxylesterase 2 (CES2). In conclusion, OM is bioactivated mainly via intestinal and additionally hepatic CMBL not only in humans but also in mice, rats, and monkeys, while CC is bioactivated via hepatic CES1 rather than intestinal enzymes, including CES2. AM is a substrate for CMBL.
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells