Merck
Ion mobility spectrometry for detection of skin volatiles.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-12-12)
Veronika Ruzsanyi, Pawel Mochalski, Alex Schmid, Helmut Wiesenhofer, Martin Klieber, Hartmann Hinterhuber, Anton Amann
ABSTRACT
Volatile organic compounds (VOCs) released by humans through their skin were investigated in near real time using ion mobility spectrometry after gas chromatographic separation with a short multi-capillary column. VOCs typically found in a small nitrogen flow covering the skin are 3-methyl-2-butenal, 6-methylhept-5-en-2-one, sec-butyl acetate, benzaldehyde, octanal, 2-ethylhexanol, nonanal and decanal at volume fractions in the low part per billion-(ppb) range. The technique presented here may contribute to elucidating some physiological processes occurring in the human skin.
Sigma-Aldrich
sec-Butyl acetate, 99%