Merck
  • Cryptophanols, new versatile compounds for the synthesis of functionalized cryptophanes and polycryptophanes.
Cryptophanols, new versatile compounds for the synthesis of functionalized cryptophanes and polycryptophanes.
Chemical communications (Cambridge, England) (2002-07-18)
Magali Darzac, Thierry Brotin, Denis Bouchu, Jean-Pierre Dutasta
ABSTRACT
After deprotection with a palladium catalyst, mono-allylated cryptophane-A (1, 2) and cryptophane-E (3) gave the new cryptophanols 4, 5 and 6, respectively, which are important key compounds for the preparation of monofunctionalized cryptophanes as well as for the design of large supramolecular hosts such as the bis-cryptophanes 7 and 8.
Sigma-Aldrich
1,10-Diiododecane, 95%
Pricing and availability is not currently available.