Merck
  • Arachidonic acid activates NADPH oxidase by a direct, calmodulin-regulated mechanism.
Arachidonic acid activates NADPH oxidase by a direct, calmodulin-regulated mechanism.
Prostaglandins & other lipid mediators (1998-07-23)
P J Hartfield, J M Robinson
ABSTRACT
In intact neutrophils, arachidonic acid rapidly and transiently activated NADPH oxidase-mediated superoxide generation. Inhibitors of protein kinases (staurosporine and H-7) failed to markedly attenuate arachidonic acid-stimulated superoxide generation. Conversely, the calmodulin antagonist W-7 blocked this arachidonic acid-stimulated response. Similarly, diphenylene iodonium potently inhibited superoxide release. These results suggest that arachidonic acid directly activates the membrane-associated NADPH oxidase and calmodulin and/or calmodulin-binding proteins are required for the assembly of the active oxidase.
Sigma-Aldrich
N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride, ≥98% (HPLC)