Merck
  • Structural study of isoflavonoids possessing antioxidant activity isolated from the fermentation broth of Streptomyces sp.
Structural study of isoflavonoids possessing antioxidant activity isolated from the fermentation broth of Streptomyces sp.
The Journal of antibiotics (1989-09-01)
S Funayama, Y Anraku, A Mita, K Komiyama, S Omura
ABSTRACT
Structures of three antioxidant isoflavonoids isolated from the cultured broth of Streptomyces sp. OH-1049 were shown to be 4',7,8-trihydroxyisoflavone (1), 3',4',7-trihydroxyisoflavone (2) and 8-chloro-3',4',5,7-tetrahydroxyisoflavone (3), respectively. Among them, 3 is a novel isoflavonoid possessing a chlorine atom in the molecule. Compound 1 was synthesized and its antitumor activities were tested against IMC carcinoma, S180, P388 leukemia and P388/ADM leukemia in vivo. As a result, 1 showed 139% increase in life span (ILS) against S180 bearing mice whereas it showed slight or no ILS against IMC carcinoma, P388 leukemia and P388/ADM leukemia bearing mice.
Sigma-Aldrich
YN1, ≥98% (HPLC)
Pricing and availability is not currently available.