Merck
  • Repeated central administration of selegiline attenuated morphine physical dependence in rat.
Repeated central administration of selegiline attenuated morphine physical dependence in rat.
Pharmacological reports : PR (2013-08-21)
Alireza Parvizpour, Mohammad Charkhpour, Bohlool Habibi-asl, Mobin Shakhsi, Majid Ghaderi, Kambiz Hassanzadeh
ABSTRACT
Long-term exposure to opiates induces physical dependence; however, the neurobiological mechanisms of this phenomenon are not completely clear. The purpose of this study was to evaluate the effects of systemic and intracerebroventricular (icv) administration of selegiline (a selective inhibitor of monoamine oxidase B) on the morphine withdrawal syndrome in rats. To this aim, adult male Sprague Dawley rats were selected randomly, and then growing doses of morphine were administered subcutaneously at an interval of 12 h for nine days with the intention of inducing dependency. Nine days after, only the morning dose of morphine was administered, followed by systemic or central injection of saline or selegiline. Later, naloxone was injected after 30 min and withdrawal signs recorded for a period of 60 min. Results showed failure of systemic administration of selegiline in changing the withdrawal symptoms; nevertheless, icv injection attenuated the withdrawal signs significantly. In conclusion we found that central administration of selegiline attenuated morphine withdrawal symptoms.