Merck
  • Study of dynamics and crystallization kinetics of 5-methyl-2-[(2-nitrophenyl)amino]-3-thiophenecarbonitrile at ambient and elevated pressure.
Study of dynamics and crystallization kinetics of 5-methyl-2-[(2-nitrophenyl)amino]-3-thiophenecarbonitrile at ambient and elevated pressure.
The Journal of chemical physics (2012-07-12)
K Adrjanowicz, K Kaminski, M Paluch, K L Ngai, Lian Yu
ABSTRACT
The organic liquid ROY, i.e., 5-methyl-2-[(2-nitrophenyl)amino]-3-thiophenecarbonitrile, has been a subject of detailed study in the last few years. One interest in ROY lies in its polymorph-dependent fast crystal growth mode below and above the glass transition temperature. This growth mode is not diffusion controlled, and the possibility that it is enabled by secondary relaxation had been suggested. However, a previous study by dielectric relaxation spectroscopy had not been able to find any resolved secondary relaxation. The present paper reports new dielectric measurements of ROY in the liquid and glassy states at ambient pressure and elevated pressure, which were performed to provide more insight into the molecular dynamics as well as the crystallization tendency of ROY. In the search of secondary relaxation, a special glassy state of ROY was prepared by applying high pressure to the liquid state, from which secondary relaxation was possibly resolved. Thus, the role of secondary relaxation in crystallization of ROY remains to be clarified. Notwithstanding, the secondary relaxation present is not necessarily the sole enabler of crystallization. In an effort to search for possible cause of crystallization other than secondary relaxation, we also performed crystallization kinetics studies of ROY at different T and P combinations while keeping the structural relaxation time constant. The results show that crystallization of ROY speeds up with pressure, opposite to the trend found in the crystallization of ibuprofen studied up to 1 GPa. The dielectric relaxation and thermodynamic properties of ROY with phenolphthalein dimethylether (PDE) are similar in many respects, but PDE does not crystallize. Taking all the above into account, besides the secondary relaxation, the specific chemical structure, molecular interactions and packing of the molecules are additional factors that could affect the kinetics of crystallization found in ROY.