Merck
  • Split-dose picosulfate, magnesium oxide, and citric acid solution markedly enhances colon cleansing before colonoscopy: a randomized, controlled trial.
Split-dose picosulfate, magnesium oxide, and citric acid solution markedly enhances colon cleansing before colonoscopy: a randomized, controlled trial.
Gastrointestinal endoscopy (2011-12-24)
Jennifer A Flemming, Stephen J Vanner, Lawrence C Hookey
ABSTRACT
Picosulfate, magnesium oxide, and citric acid solution is a small-volume agent for colon cleansing before colonoscopy that is extremely well tolerated by patients, safe, and efficacious. Studies of other cleansing agents have suggested that split-dose regimens may further enhance efficacy. To examine whether split-dosing of picosulfate, magnesium oxide, and citric acid solution increases bowel cleansing efficacy while maintaining tolerability and safety. Prospective, randomized, single-blinded, controlled trial. Outpatient tertiary care center. A total of 236 patients underwent colonoscopy (mean age 56 years, 53.8% female). Patients in the traditional arm (n = 123) consumed 1 sachet of solution at 5:00 pm and 10:00 pm the night before the colonoscopy. Patients in the split-dose arm (n = 127) consumed 1 sachet at 7:00 pm the night before and another sachet 4 hours before their colonoscopy appointment. Ottawa Bowel Preparation Scale (OBPS) score, Aronchick score, safety, tolerability. The 113 and 109 patients in the split-dose and traditional arms, respectively, had OBPS scores for analysis. Overall, the OBPS scores in the split-dose group were significantly improved compared with the traditional dose group (4.05 vs 5.51, P < .001). This was mostly attributed to improvements in right-sided colon cleansing (1.22 in split-dose vs 2.14 in traditional arm, P < .001). Both regimens were well tolerated by patients, and no safety issues were identified. This was a single-center study. Disturbances in sleep related to the preparation were not assessed. The split-dose regimen of picosulfate, magnesium oxide, and citric acid solution is superior to the traditional dosing regimen for colon cleansing before colonoscopy. ( NCT00885274.).
Product Description
Picosulfate for system suitability, European Pharmacopoeia (EP) Reference Standard