Merck
  • Biotechnological production of 20-alpha-dihydrodydrogesterone at pilot scale.
Biotechnological production of 20-alpha-dihydrodydrogesterone at pilot scale.
Applied biochemistry and biotechnology (2011-04-16)
Julia Maria Naumann, Andy Zöllner, Călin-Aurel Drăgan, Josef Messinger, Joachim Adam, Matthias Bureik
ABSTRACT
The human sex hormone progesterone plays an essential and complex role in a number of physiological processes. Progesterone deficiency is associated with menstrual disorders and infertility as well as premature birth and abortion. For progesterone replacement therapy, the synthetic progestogen dydrogesterone is commonly used. In the body, this drug is metabolized to 20α-dihydrodydrogesterone (20α-DHD), which also shows extensive pharmacological effects and hence could act as a therapeutic agent itself. In this study, we describe an efficient biotechnological production procedure for 20α-DHD that employs the stereo- and regioselective reduction of dydrogesterone in a whole-cell biotransformation process based on recombinant fission yeast cells expressing the human enzyme AKR1C1 (20α-hydroxysteroid dehydrogenase, 20α-HSD). In a fed-batch fermentation at pilot scale (70 L) with a genetically improved production strain and under optimized reaction conditions, an average 20α-DHD production rate of 190 μM day(-1) was determined for a total biotransformation time of 136 h. Combined with an effective and reliable downstream processing, a continuous production rate of 12.3 ± 1.4 g 20α-DHD per week and fermenter was achieved. We thus established an AKR-dependent whole-cell biotransformation process that can also be used for the production of other AKR1C1 substrates (as exemplarily shown by the production of 20α-dihydroprogesterone in gram scale) and is in principle suited for the production of further human AKR metabolites at industrial scale.
Product Description
Dydrogesterone impurity B, European Pharmacopoeia (EP) Reference Standard
Dydrogesterone, European Pharmacopoeia (EP) Reference Standard