Merck
  • Regioselectivity in beta-galactosidase-catalyzed transglycosylation for the enzymatic assembly of D-galactosyl-D-mannose.
Regioselectivity in beta-galactosidase-catalyzed transglycosylation for the enzymatic assembly of D-galactosyl-D-mannose.
Bioscience, biotechnology, and biochemistry (2004-10-27)
Mariko Miyasato, Katsumi Ajisaka
ABSTRACT
The regioselectivity of beta-galactosidase derived from Bacillus circulans ATCC 31382 (beta-1,3-galactosidase) in transgalactosylation reactions using D-mannose as an acceptor was investigated. This D-mannose associated regioselectivity was found to be different from reactions using either GlcNAc or GalNAc as acceptors, not only for beta-1,3-galactosidase but also for beta-galactosidases of different origins. The relative hydrolysis rate of Gal beta-pNP and D-galactosyl-D-mannoses, of various linkages, was also measured in the presence of beta-1,3-galactosidase and was found to correlate well with the ratio of disaccharides formed by transglycosylation. The unexpected regioselectivity using D-mannose can therefore be explained by an anomalous specificity in the hydrolysis reaction. By utilizing the identified characteristics of both regioselectivity and hydrolysis specificity using D-mannose, an efficient method for enzymatic synthesis of beta-1,3-, beta-1,4- and beta-1,6-linked D-galactosyl-D-mannose was subsequently established.