Merck
  • Pharmacokinetics of 17 beta-dihydroequilin sulfate in normal postmenopausal women under steady state conditions.
Pharmacokinetics of 17 beta-dihydroequilin sulfate in normal postmenopausal women under steady state conditions.
Journal of the Society for Gynecologic Investigation (2002-10-26)
Bhagu R Bhavnani, Anthony Cecutti, Alan Gerulath
ABSTRACT
In the present study, the constant infusion of [3H]17 beta-dihydroequilin sulfate ([3H]17 beta-EqS) was used to estimate the metabolic clearance rate (MCR) of 17 beta-dihydroequilin sulfate (17 beta-EqS) and to measure the conversion of this estrogen to equilin sulfate (EqS), equilenin sulfate (EqnS), 17 beta-dihydroequilenin sulfate (17 beta-EqnS), equilin (Eq), equilenin (Eqn), 17 beta-dihydroequilin (17 beta-Eq), and 17 beta-dihydroequilenin (17 beta-Eqn) in normal postmenopausal women. In seven healthy postmenopausal women, infusion of [3H]17 beta-EqS was started 30 minutes after a priming dose and continued at a constant rate of 10-20 microCi/hour, for 3-6 hours. Three blood samples were taken during and at the end of infusion. From the plasma, unconjugated and sulfate-conjugated estrogens, 17 beta-EqS, EqS, EqnS, 17 beta-EqnS, Eq, Eqn, 17 beta-Eq, and 17 beta-Eqn were isolated and purified by high performance liquid chromatography. The MCR of 17 beta-EqS and the conversion ratios and transfer constants (rho) for precursor (17 beta-EqS) to products were calculated. The mean MCR of 17 beta-EqS was calculated to be 797 +/- 90 L/day or 506 +/- 60 L/m2 per day. The mean conversion ratio (CRPRE-PROBB) was 2.4 +/- 0.4 for EqS, 0.3 +/- 0.04 for EqnS, 0.25 +/- 0.03 for 17 beta-EqnS, 0.09 +/- 0.02 for Eq, 0.03 +/- 0.01 for Eqn, 0.08 +/- 0.02 for 17 beta-Eq, and 0.03 +/- 0.01 for 17 beta-Eqn. In both the sulfate-conjugated and unconjugated forms, the most abundant metabolite formed was Eq. Based on the previously reported MCR of EqS (170 L/m2 per day) and 17 beta-Eq (1252 L/m2 per day), the transfer constants [rho]BB were calculated to be 0.8 +/- 0.10 and 0.20 +/- 0.03, respectively. The results indicate that a large portion of 17 beta-EqS is converted to EqS and the more potent estrogen 17 beta-Eq. The ratio of rho EqS-17 beta-EqS to rho 17 beta-EqS-EqS was calculated to be 0.8 +/- 0.1 and represents the extent of C-17-oxidation and reduction and indicates that substantial amounts of 17 beta-reduced metabolites will still be present in the blood although the oxidation reaction was somewhat greater. The data indicate that, compared with the classic estrogens, the in vivo metabolism of ring B unsaturated estrogens is complex. Thus, although the amount of 17 beta-EqS originally present in the conjugated equine estrogens is small, the pharmacokinetics and pharmacodynamics of EqS, 17 beta-EqS, and the extensive interconversions between these estrogens support the hypothesis that the major in vivo activity of the EqS present in conjugated equine estrogens is expressed through its metabolism to 17 beta-EqS and 17 beta-Eq. Furthermore, the increased estrogenic activity associated with this drug may in part be due to the formation of these 17 beta-reduced metabolites.
Product Description
17α-Dihydroequilin, European Pharmacopoeia (EP) Reference Standard