Merck
Resolution and absolute configuration of naturally occurring auronols.
Journal of natural products (2001-03-30)
R Bekker, X C Li, H N ElSohly, A M Clark, E V Brandt, D Ferreira
ABSTRACT
Resolution of racemic 2-benzyl-2-hydroxy-1-benzofuran-3(2H)-ones (auronols) and CD data of the ensuing enantiomers permit assessment of the absolute configuration at the single stereocenter.
Sigma-Aldrich
Maesopsin, ≥90% (LC/MS-ELSD)
Pricing and availability is not currently available.