- GAPDH Pseudogenes and the Quantification of Feline Genomic DNA Equivalents.
GAPDH Pseudogenes and the Quantification of Feline Genomic DNA Equivalents.
Quantitative real-time PCR (qPCR) is broadly used to detect and quantify nucleic acid targets. In order to determine cell copy number and genome equivalents, a suitable reference gene that is present in a defined number in the genome is needed, preferably as a single copy gene. For most organisms, a variable number of glyceraldehyde-3-phosphate dehydrogenase (GAPDH) pseudogenes have been reported. However, it has been suggested that a single-copy of the GAPDH pseudogene is present in the feline genome and that a GAPDH assay can therefore be used to quantify feline genomic DNA (gDNA). The aim of this study was to determine whether one or more GAPDH pseudogenes are present in the feline genome and to provide a suitable alternative qPCR system for the quantification of feline cell copy number and genome equivalents. Bioinformatics and sequencing results revealed that not just one but several closely related GAPDH-like sequences were present in the cat genome. We thus identified, developed, optimized, and validated an alternative reference gene assay using feline albumin (fALB). Our data emphasize the need for an alternative reference gene, apart from the GAPDH pseudogene, for the normalization of gDNA levels. We recommend using the fALB qPCR assay for future studies.