Merck
  • A one-pot access to 6-substituted phenanthridines from fluoroarenes and nitriles via 1,2-arynes.
A one-pot access to 6-substituted phenanthridines from fluoroarenes and nitriles via 1,2-arynes.
Organic letters (2002-08-03)
Jan Pawlas, Mikael Begtrup
ABSTRACT
[reaction: see text] A one-pot, t-BuLi-induced synthesis of 6-substituted phenanthridines from fluoroarenes and nitriles via 1,2-arynes is reported. Aryl- and hetaryl nitriles, cyanamides, and trimethylacetonitrile gave phenanthridine products. The method was extended to provide bisphenanthridine 10 by a one-pot bis-cyclization, using 1,3-dicyanobenzene and PhF in 1:5 ratio. Reaction of 1-fluoronaphthalene and 4-chlorofluorobenzene with benzonitrile afforded the regioisomerically pure products 11 and 12, respectively.
Sigma-Aldrich
1-Fluoronaphthalene, 99%
Pricing and availability is not currently available.