Merck
  • [Preparation of cinnamomi cortex oil microspheres based on porous silicon dioxide and its property characterizations].
[Preparation of cinnamomi cortex oil microspheres based on porous silicon dioxide and its property characterizations].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2014-02-05)
Chun-Xia Zhu, Yan-Rong Jiang, Zhen-Hai Zhang, Dong-Mei Ding, Xiao-Bin Jia
ABSTRACT
To determine the optimum process for preparing Cinnamomi Cortex oil microspheres based on porous silicon dioxide. After porous silica dioxide adsorbed Cinnamomi Cortex oil, Cinnamomi Cortex oil microspheres were prepared by the dropping method, with sodium alginate as the skeleton materials. The preparation process was optimized through the L(9) (3(4)) orthogonal test design, with microspheres diameter, distribution, drug loading capacity and entrapment efficiency as the indexes. The cinnamon volatile oil microspheres were characterized by scanning election microscope (SEM), thermogravimetric analysis (TGA), and infrared (IR) spectroscopy. An in vitro drug release experiment was conducted. The results showed that the microspheres prepared with the optimal process parameters were in good shape, even in size and good in dispersibility, with an average diameter of 1.61 mm, an average drug loading capacity of 32.85%, an entrapment efficiency of 94.79%. The maximum drug release capacity reached 72.6%, 95.0%, 97.4%, respectively, under pH 4.0, 6.8, 7.4 in 6 hours. Meanwhile, microsphere generation was tested by IR, TGA and other methods. The established optimum process for preparing Cinnamomi Cortex oil microspheres was proved to be stable and practical.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Alginic acid sodium salt, powder
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, ≥99.9% trace metals basis
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Sodium alginate
Sigma-Aldrich
Alginic acid from brown algae, powder