Merck
  • Role of diuretics, β blockers, and statins in increasing the risk of diabetes in patients with impaired glucose tolerance: reanalysis of data from the NAVIGATOR study.
Role of diuretics, β blockers, and statins in increasing the risk of diabetes in patients with impaired glucose tolerance: reanalysis of data from the NAVIGATOR study.
BMJ (Clinical research ed.) (2013-12-11)
Lan Shen, Bimal R Shah, Eric M Reyes, Laine Thomas, Daniel Wojdyla, Peter Diem, Lawrence A Leiter, Bernard Charbonnel, Viacheslav Mareev, Edward S Horton, Steven M Haffner, Vladimir Soska, Rury Holman, M Angelyn Bethel, Frank Schaper, Jie-Lena Sun, John J V McMurray, Robert M Califf, Henry Krum
ABSTRACT
To examine the degree to which use of β blockers, statins, and diuretics in patients with impaired glucose tolerance and other cardiovascular risk factors is associated with new onset diabetes. Reanalysis of data from the Nateglinide and Valsartan in Impaired Glucose Tolerance Outcomes Research (NAVIGATOR) trial. NAVIGATOR trial. Patients who at baseline (enrolment) were treatment naïve to β blockers (n=5640), diuretics (n=6346), statins (n=6146), and calcium channel blockers (n=6294). Use of calcium channel blocker was used as a metabolically neutral control. Development of new onset diabetes diagnosed by standard plasma glucose level in all participants and confirmed with glucose tolerance testing within 12 weeks after the increased glucose value was recorded. The relation between each treatment and new onset diabetes was evaluated using marginal structural models for causal inference, to account for time dependent confounding in treatment assignment. During the median five years of follow-up, β blockers were started in 915 (16.2%) patients, diuretics in 1316 (20.7%), statins in 1353 (22.0%), and calcium channel blockers in 1171 (18.6%). After adjusting for baseline characteristics and time varying confounders, diuretics and statins were both associated with an increased risk of new onset diabetes (hazard ratio 1.23, 95% confidence interval 1.06 to 1.44, and 1.32, 1.14 to 1.48, respectively), whereas β blockers and calcium channel blockers were not associated with new onset diabetes (1.10, 0.92 to 1.31, and 0.95, 0.79 to 1.13, respectively). Among people with impaired glucose tolerance and other cardiovascular risk factors and with serial glucose measurements, diuretics and statins were associated with an increased risk of new onset diabetes, whereas the effect of β blockers was non-significant. ClinicalTrials.gov NCT00097786.
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
SAFC
L-Valine
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Valsartan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Valsartan, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Nateglinide, ≥98% (HPLC), solid
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Valsartan for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
N-[(cis-4-Isopropylcyclohexyl)carbonyl]-D-phenylalanine, European Pharmacopoeia (EP) Reference Standard