Merck
  • A circadian clock gene, Rev-erbα, modulates the inflammatory function of macrophages through the negative regulation of Ccl2 expression.
A circadian clock gene, Rev-erbα, modulates the inflammatory function of macrophages through the negative regulation of Ccl2 expression.
Journal of immunology (Baltimore, Md. : 1950) (2013-12-07)
Shogo Sato, Takuya Sakurai, Junetsu Ogasawara, Motoko Takahashi, Tetsuya Izawa, Kazuhiko Imaizumi, Naoyuki Taniguchi, Hideki Ohno, Takako Kizaki
ABSTRACT
Disruption of the circadian rhythm is a contributory factor to clinical and pathophysiological conditions, including cancer, the metabolic syndrome, and inflammation. Chronic and systemic inflammation are a potential trigger of type 2 diabetes and cardiovascular disease and are caused by the infiltration of large numbers of inflammatory macrophages into tissue. Although recent studies identified the circadian clock gene Rev-erbα, a member of the orphan nuclear receptors, as a key mediator between clockwork and inflammation, the molecular mechanism remains unknown. In this study, we demonstrate that Rev-erbα modulates the inflammatory function of macrophages through the direct regulation of Ccl2 expression. Clinical conditions associated with chronic and systemic inflammation, such as aging or obesity, dampened Rev-erbα gene expression in peritoneal macrophages from C57BL/6J mice. Rev-erbα agonists or overexpression of Rev-erbα in the murine macrophage cell line RAW264 suppressed the induction of Ccl2 following an LPS endotoxin challenge. We discovered that Rev-erbα represses Ccl2 expression directly through a Rev-erbα-binding motif in the Ccl2 promoter region. Rev-erbα also suppressed CCL2-activated signals, ERK and p38, which was recovered by the addition of exogenous CCL2. Further, Rev-erbα impaired cell adhesion and migration, which are inflammatory responses activated through the ERK- and p38-signaling pathways, respectively. Peritoneal macrophages from mice lacking Rev-erbα display increases in Ccl2 expression. These data suggest that Rev-erbα regulates the inflammatory infiltration of macrophages through the suppression of Ccl2 expression. Therefore, Rev-erbα may be a key link between aging- or obesity-associated impairment of clockwork and inflammation.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine 1 M solution
Sigma-Aldrich
Anti-NR1D1 (Rev-ErbA α) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)