Merck
  • Simultaneous determination of 13-cis and all-trans vitamin A palmitate (retinyl palmitate), vitamin A acetate (retinyl acetate), and total vitamin E (alpha-tocopherol and DL-alpha-tocopherol acetate) in infant formula and adult nutritionals by normal phase HPLC: first action 2012.10.
Simultaneous determination of 13-cis and all-trans vitamin A palmitate (retinyl palmitate), vitamin A acetate (retinyl acetate), and total vitamin E (alpha-tocopherol and DL-alpha-tocopherol acetate) in infant formula and adult nutritionals by normal phase HPLC: first action 2012.10.
Journal of AOAC International (2013-11-29)
Adrienne McMahon, Scott Christiansen, Lynsey Shine, Calvin Loi, Dawn Dowell
ABSTRACT
This HPLC method, with both variable UV and fluorescence detection, allows for the simultaneous determination of vitamin A palmitate, vitamin A acetate, and total vitamin E in infant, pediatric, and adult nutritional formulas. The concentration of each vitamin form is calculated by comparison with standards of known concentration. Following hydrolysis, the vitamins are extracted into iso-octane and analyzed by normal phase (NP) HPLC. The method was evaluated for linearity, precision, and accuracy using a selection of the Stakeholder Panel on Infant Formula and Adult Nutritionals (SPIFAN) matrixes, including milk-based, soy-based, and hydrolyzed protein, as well as high- and low-fat products. A single-laboratory validation has been completed for all analytes using a selection of SPIFAN matrixes. Performance parameters included a working range of 2-450 microg/100 g ready-to-feed for vitamin A and 0.03-8.0 mgl100 g reconstituted final product for vitamin E. LOD was <1.0 microg and <0.1 mgl100 g reconstituted final product for vitamins A and E, respectively; RSD was 1.08-8.70% over a range of concentration; and average recoveries of 97.4-101.3%. Repeatability of <4% for vitamin A and <8% for vitamin E was calculated from five laboratories using this method. Results indicate that this method is suitable for the analysis of vitamins A and E in all forms of infant, adult, and pediatric formulas (powders, ready-to-feed liquids, and liquid concentrates). The Expert Review Panel (ERP) of Infant Formula reviewed this method separately for vitamins A and E, including all available method validation data at the AOAC INTERNATIONAL Annual Meeting on September 29, 2012. Following evaluation of the data for both methods, the ERP agreed that both methods met the standard method performance requirements articulated by SPIFAN. The ERP granted First Action status to both methods, and recommended that a single method be published for the simultaneous determination of vitamin A palmitate, vitamin A acetate, and total vitamin E (DL-a-tocopherol and DL-alpha-tocopherol acetate) in infant formula and adult nutritionals by NP HPLC.
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil
Sigma-Aldrich
Retinyl palmitate, potency: ≥1,700,000 USP units per g
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Supelco
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Retinol palmitate, analytical standard
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
Sigma-Aldrich
Retinyl acetate, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g
Retinol acetate, European Pharmacopoeia (EP) Reference Standard
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material