Merck
In vitro to in vivo profiling: an easy idea for biowaiver study.
Acta poloniae pharmaceutica (2013-10-24)
Abdulhakim A A Khaled, Khalid Pervaiz, Sonia Khiljee, Sabiha Karim, Qurat-ul-Ain Shoaib, Ghulam Murtaza
ABSTRACT
The aim of this article was to assess and apply the in vitro to in vivo profiling (IVIVP), a new biowaiver approach, in designing a product with specific release pattern. The IVIVP was established by plotting the observed and predicted plasma drug concentrations. For IVIVP, convolution approach was employed to estimate plasma drug concentrations from in vitro dissolution profiles. The IVIVP for T1S exhibited a good correlation coefficient (R2 = 0.963) followed by the T2 (R2 = 0.682), T3 (R2 = 0.665), T1 (R2 = 0.616), and Mepresso (R2 = 0.345). Establishing an IVIVP, based on the convolution approach, can be more useful and practicable in the biowaiver studies, rather than present not useful practice of IVIVC estimated via deconvolution approach. This paper also elaborates that there is good correlation between the in vitro and in vivo profiles of the developed metoprolol tartrate formulations, particularly for T1S.
Sigma-Aldrich
(±)-Metoprolol (+)-tartrate salt, ≥98% (titration), powder
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Sigma-Aldrich
Poly(methyl methacrylate-co-methacrylic acid), average Mw ~34,000 by GPC, average Mn ~15,000 by GPC
Metoprolol tartrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Metoprolol Tartrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Metoprolol tartrate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®