Merck
  • Conjugated linoleate reduces prostate cancer viability whereas the effects of oleate and stearate are cell line-dependent.
Conjugated linoleate reduces prostate cancer viability whereas the effects of oleate and stearate are cell line-dependent.
Anticancer research (2013-10-15)
Rachel M Hagen, Anthony Rhodes, Michael R Ladomery
ABSTRACT
In this study, responses to fatty acid treatments in commonly used prostate cancer cell culture models and variability of gene expression between them were determined. PC3, DU145, LNCaP, VCaP and PNT2 cells were treated with 100 μM of either oleate, stearate or conjugated linoleate. Cell proliferation and viability were assessed using trypan blue and 3-(4, 5-dimethylthiazol-2-yl)-2, 5-diphenyltetrazolium bromide (MTT) assay respectively. Gene expression was measured using real-time polymerase chain reaction (PCR). Conjugated linoleic acid reduced cell proliferation and viability in all prostate cancer cell lines, whilst the effects of oleic and stearic acid on proliferation were found to be cell line-dependent. A reduction in gene expression of fatty acid desaturases was observed in prostate cancer cell lines compared to normal prostate cells. Differential responses of the cell lines investigated here to fatty acid treatment suggest that multiple prostate cancer cell line models should be used when designing experiments aimed at examining lipid metabolism in prostate cancer.
Sigma-Aldrich
Acetyl-CoA carboxylase 2 human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Oleic acid, analytical standard
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, natural, FCC
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetyl-CoA Carboxylase 1 human, recombinant, expressed in Sf9 cells
Supelco
Oleic acid, Selectophore, ≥99%