Merck
  • Occurrence of phthalic acid esters in source waters: a nationwide survey in China during the period of 2009-2012.
Occurrence of phthalic acid esters in source waters: a nationwide survey in China during the period of 2009-2012.
Environmental pollution (Barking, Essex : 1987) (2013-10-01)
Xiaowei Liu, Jianghong Shi, Ting Bo, Hui Zhang, Wei Wu, Qingcai Chen, Xinmin Zhan
ABSTRACT
The first nationwide survey of six phthalic acid esters (PAEs) (diethyl phthalate (DEP); dimethyl phthalate (DMP); di-n-butyl phthalate (DBP); butyl benzyl phthalate (BBP); bis(2-ethylhexyl) phthalate (DEHP); di-n-octyl phthalate (DnOP)) in source waters was conducted in China. The results showed these PAEs were ubiquitous in source waters. DBP and DEHP were the most frequently detected with high concentrations ranging nd-1.52 μg/L and nd-6.35 μg/L, respectively. These PAEs concentrations (except DBP) in surface water (rivers, lakes and reservoirs) were generally higher than those in groundwater; DBP had high concentrations in groundwater in Northeast China (Liao River Basin) and North China (Hai River Basin). Their concentrations in the northern regions were generally higher than those in the southern and eastern regions; particularly, in North China. Three short-chain PAEs (DMP, DEP and DBP) were detected with high concentrations in Hai River Basin, Pearl River Basin and Yellow River Basin.
Product Description
Supelco
Benzyl butyl phthalate, analytical standard
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzyl butyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzyl butyl phthalate, 98%
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Dimethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Supelco
Di-n-butyl phthalate solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Di-n-octyl phthalate, analytical standard
Sigma-Aldrich
Diethyl phthalate, ≥99%
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Supelco
Diethyl phthalate, PESTANAL®, analytical standard
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Di-n-octyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Sigma-Aldrich
Di-n-octyl phthalate, ≥98.0% (GC)
Sigma-Aldrich
Diethyl phthalate, 99.5%
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)